Learn More
2,2'-Bithiophene, 97%, Thermo Scientific Chemicals
£45.10 - £205.00
Chemical Identifiers
CAS | 492-97-7 |
---|---|
Molecular Formula | C8H6S2 |
Molecular Weight (g/mol) | 166.27 |
MDL Number | MFCD00005414 |
InChI Key | OHZAHWOAMVVGEL-UHFFFAOYSA-N |
Synonym | 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene |
PubChem CID | 68120 |
ChEBI | CHEBI:36821 |
IUPAC Name | 2-thiophen-2-ylthiophene |
SMILES | C1=CSC(=C1)C2=CC=CS2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10117420
|
Thermo Scientific Acros
402690025 |
2.5 g | Glass bottle |
£45.10
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10289830
|
Thermo Scientific Acros
402690200 |
20 g | Glass bottle |
£205.00
20g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
492-97-7 | |
166.27 | |
OHZAHWOAMVVGEL-UHFFFAOYSA-N | |
68120 | |
2-thiophen-2-ylthiophene |
C8H6S2 | |
MFCD00005414 | |
2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene | |
CHEBI:36821 | |
C1=CSC(=C1)C2=CC=CS2 |
Specifications
492-97-7 | |
100.0 | |
260.0°C | |
96% min. (GC) | |
MFCD00005414 | |
2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene | |
OHZAHWOAMVVGEL-UHFFFAOYSA-N | |
2-thiophen-2-ylthiophene | |
68120 | |
166.27 | |
2, 2'-Bithiophene, 97% |
96.0 | |
30.0°C to 35.0°C | |
>110°C | |
C8H6S2 | |
19, 32 | |
Solubility in water: slightly soluble. Other solubilities: soluble in methanol | |
C1=CSC(=C1)C2=CC=CS2 | |
166.27 | |
CHEBI:36821 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-767-2
missing translation for 'tsca' : TSCA
- 2,2'-Bithiophene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only