Learn More
2-(2-Bromoethyl)-1,3-dioxane, 98%, Thermo Scientific Chemicals
£29.90 - £109.00
Chemical Identifiers
CAS | 33884-43-4 |
---|---|
Molecular Formula | C6H11BrO2 |
Molecular Weight (g/mol) | 195.06 |
MDL Number | MFCD00006567 |
InChI Key | KXMZOKKPQZRPRN-UHFFFAOYNA-N |
Synonym | 2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane |
PubChem CID | 520656 |
IUPAC Name | 2-(2-bromoethyl)-1,3-dioxane |
SMILES | CC(Br)C1OCCCO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10677030
|
Thermo Scientific Acros
297930100 |
10 g | Glass bottle |
£29.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10461050
|
Thermo Scientific Acros
297930500 |
50 g | Glass bottle |
£109.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33884-43-4 | |
195.06 | |
KXMZOKKPQZRPRN-UHFFFAOYNA-N | |
520656 | |
CC(Br)C1OCCCO1 |
C6H11BrO2 | |
MFCD00006567 | |
2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane | |
2-(2-bromoethyl)-1,3-dioxane |
Specifications
33884-43-4 | |
100.0 | |
1.4400g/mL | |
97°C | |
97.5% min. (GC) | |
MFCD00006567 | |
2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane | |
CC(Br)C1OCCCO1 | |
195.06 | |
195.06 | |
Liquid |
97.5 | |
Colorless | |
67.0°C to 70.0°C (2.8 mmHg) | |
Authentic | |
C6H11BrO2 | |
1.44 | |
KXMZOKKPQZRPRN-UHFFFAOYNA-N | |
2-(2-bromoethyl)-1,3-dioxane | |
520656 | |
98% | |
2-(2-Bromoethyl)-1,3-dioxane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 251-716-7
- 2-(2-Bromoethyl)-1,3-dioxane
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P362-Take off contaminated clothing.
RUO – Research Use Only