Learn More
2,2-Di(tert-butylperoxy)butane, 50% solution in aromatic free mineral spirit, Thermo Scientific Chemicals
£14.07 - £46.83
Chemical Identifiers
CAS | 2167-23-9, 31807-55-3, 78-93-3 |
---|---|
Molecular Formula | C12H26O4 |
Molecular Weight (g/mol) | 234.336 |
MDL Number | MFCD00048243 |
InChI Key | HQOVXPHOJANJBR-UHFFFAOYSA-N |
Synonym | 2,2-di tert-butylperoxy butane, 2,2-bis tert-butylperoxy butane, chaloxyd p 1293al, trigonox d, chaloxyd p 1200al, butane, 2,2-bis tert-butyldioxy, unii-v15k90hftj, v15k90hftj, 2,2-bis t-butylperoxy butane, di-tert-butyl sec-butylidene diperoxideShow More |
PubChem CID | 221260 |
IUPAC Name | 2,2-bis(tert-butylperoxy)butane |
SMILES | CCC(C)(OOC(C)(C)C)OOC(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10673892
|
Thermo Scientific Acros
349830100 |
10 mL | Plastic bottle |
£14.07
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10499252
|
Thermo Scientific Acros
349835000 |
500 mL | Plastic bottle |
£46.83
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsChemical Identifiers
2167-23-9, 31807-55-3, 78-93-3 | |
234.336 | |
HQOVXPHOJANJBR-UHFFFAOYSA-N | |
221260 | |
CCC(C)(OOC(C)(C)C)OOC(C)(C)C |
C12H26O4 | |
MFCD00048243 | |
2,2-di tert-butylperoxy butane, 2,2-bis tert-butylperoxy butane, chaloxyd p 1293al, trigonox d, chaloxyd p 1200al, butane, 2,2-bis tert-butyldioxy, unii-v15k90hftj, v15k90hftj, 2,2-bis t-butylperoxy butane, di-tert-butyl sec-butylidene diperoxideShow More | |
2,2-bis(tert-butylperoxy)butane |
Specifications
Undesignated | |
49.0 | |
C12H26O4 | |
0.8050g/mL | |
Solubility in water: immiscible. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons | |
CCC(C)(OOC(C)(C)C)OOC(C)(C)C | |
234.336 | |
234.34 | |
0.805 | |
43°C | |
2, 2-Di(tert-butylperoxy)butane, 50% Solution in Aromatic Free Mineral Spirit |
2167-23-9, 31807-55-3, 78-93-3 | |
51.0 | |
MFCD00048243 | |
2,2-di tert-butylperoxy butane, 2,2-bis tert-butylperoxy butane, chaloxyd p 1293al, trigonox d, chaloxyd p 1200al, butane, 2,2-bis tert-butyldioxy, unii-v15k90hftj, v15k90hftj, 2,2-bis t-butylperoxy butane, di-tert-butyl sec-butylidene diperoxide | |
HQOVXPHOJANJBR-UHFFFAOYSA-N | |
2,2-bis(tert-butylperoxy)butane | |
221260 | |
49 to 51% | |
2.16 mPa.s (20°C) | |
Solution |
Safety and Handling
GHS H Statement:
Flammable liquid and vapour.
Heating may cause a fire.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
May be fatal if swallowed and enters airways.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 218-507-2
missing translation for 'tsca' : TSCA
- 2,2-Di(tert-butylperoxy)butane, 50% solution in aromatic free mineral spirit
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Organic peroxide Type C & D
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H242-Heating may cause a fire.
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.