missing translation for 'onlineSavingsMsg'
Learn More

(2,2-Dichloro-1-methylcyclopropyl)benzene, 95%, Thermo Scientific™

Product Code. 11417256
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3591-42-2
Molecular Formula C10H10Cl2
Molecular Weight (g/mol) 201.09
MDL Number MFCD00012260
InChI Key NXYPRVQXSWVEOB-UHFFFAOYSA-N
Synonym 2,2-dichloro-1-methylcyclopropyl benzene, benzene, 2,2-dichloro-1-methylcyclopropyl, 1,1-dichloro-2-methyl-2-phenylcyclopropane, 2,2-dichloro-1-methyl cyclopropyl benzene, 2,2-dichloro-1-methyl-cyclopropyl benzene, acmc-20aok2, 1-phenyl-1-methyl-2,2-dichlorocyclopropane, 1-methyl-2,2-dichlorocyclopropylbenzene, 2,2-dichloro-1-methyl-cyclopropyl-benzene, 2,2-dichloro-1-methyl-1-phenyl-cyclopropane
PubChem CID 107129
IUPAC Name (2,2-dichloro-1-methylcyclopropyl)benzene
SMILES CC1(CC1(Cl)Cl)C2=CC=CC=C2

Specifications

Boiling Point 70°C to 72°C (0.8 mmHg)
Refractive Index 1.541
Quantity 5 g
Formula Weight 201.09
Percent Purity 95%
Chemical Name or Material (2,2-Dichloro-1-methylcyclopropyl)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'