missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Thermo Scientific™
£108.80 - £537.60
Chemical Identifiers
CAS | 128073-33-6 |
---|---|
InChI Key | HLFLYOQLHYYNLT-UHFFFAOYSA-N |
Synonym | 2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid |
PubChem CID | 14695229 |
IUPAC Name | 2,2-difluoro-1-methylcyclopropane-1-carboxylic acid |
SMILES | CC1(CC1(F)F)C(=O)O |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
128073-33-6 | |
2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid | |
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid |
HLFLYOQLHYYNLT-UHFFFAOYSA-N | |
14695229 | |
CC1(CC1(F)F)C(=O)O |
Specifications
128073-33-6 | |
97% | |
2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid | |
CC1(CC1(F)F)C(=O)O | |
136.098 | |
136.1 | |
(±)-2, 2-Difluoro-1-methylcyclopropanecarboxylic acid |
62.0°C to 65.0°C | |
C5H6F2O2 | |
HLFLYOQLHYYNLT-UHFFFAOYSA-N | |
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid | |
14695229 | |
97% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.