Learn More
2,2-Dimethoxypropane, 98+%, Thermo Scientific Chemicals
£34.30 - £347.00
Chemical Identifiers
CAS | 77-76-9 |
---|---|
Molecular Formula | C5H12O2 |
Molecular Weight (g/mol) | 104.15 |
InChI Key | HEWZVZIVELJPQZ-UHFFFAOYSA-N |
Synonym | acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan |
PubChem CID | 6495 |
IUPAC Name | 2,2-dimethoxypropane |
SMILES | CC(C)(OC)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10274330
|
Thermo Scientific Acros
115635000 |
500 mL | Glass bottle |
£34.30
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10214290
|
Thermo Scientific Acros
115630010 |
1 L | Glass bottle |
£55.70
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10410384
|
Thermo Scientific Acros
115630025 |
2.5 L | Glass bottle |
£104.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11994761
|
Thermo Scientific Acros
115630100 |
10 L | Metal Drum |
£347.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
77-76-9 | |
104.15 | |
acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan | |
2,2-dimethoxypropane |
C5H12O2 | |
HEWZVZIVELJPQZ-UHFFFAOYSA-N | |
6495 | |
CC(C)(OC)OC |
Specifications
77-76-9 | |
100.0 | |
Colorless | |
83.0°C | |
Authentic | |
C5H12O2 | |
1.3760 to 1.3780 | |
01,268; 05,226; 13,199; 15,138 | |
acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan | |
HEWZVZIVELJPQZ-UHFFFAOYSA-N | |
2,2-dimethoxypropane | |
6495 | |
104.15 | |
Liquid |
98.0 | |
-47.0°C | |
0.8400g/mL | |
−5°C | |
98% min. (GC) | |
(CH3)2C(OCH3)2 | |
01,648 | |
0.84 | |
Solubility in water: 18 g/100g (25°C). Other solubilities: soluble in acetone, benzene, ccl4, ether,, n-heptane and methanol | |
CC(C)(OC)OC | |
104.15 | |
0.452 mPa.s (25°C) | |
98+% | |
2,2-Dimethoxypropane |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-056-
- 2,2-Dimethoxypropane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P310-Immediately call a POISON CENTER/doctor/.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only