missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals

Product Code. 11935191
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7328-91-8
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
InChI Key DDHUNHGZUHZNKB-UHFFFAOYSA-N
PubChem CID 81770
IUPAC Name 2,2-dimethylpropane-1,3-diamine
SMILES CC(C)(CN)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29.0°C to 31.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NCH2C(CH3)2CH2NH2
Refractive Index 1.4566
Quantity 50 g
Beilstein 04,III,595
Specific Gravity 0.85
Synonym 2,2-dimethyl-1,3-propanediamine, neopentyldiamine, 1,3-propanediamine, 2,2-dimethyl, neopentanediamine, 1,3-diamino-2,2-dimethylpropane, 2,2-dimethyltrimethylenediamine, 1,3-propanediamine,2,2-dimethyl, neopentylenediamine, 2,3-propanediamine, acmc-209oqg
Solubility Information Solubility in water: miscible
Formula Weight 102.18
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2, 2-Dimethyl-1, 3-propanediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2-Dimethyl-1,3-propanediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'