Learn More
2,2-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals
£87.23 - £281.40
Chemical Identifiers
CAS | 7328-91-8 |
---|---|
Molecular Formula | C5H14N2 |
Molecular Weight (g/mol) | 102.18 |
InChI Key | DDHUNHGZUHZNKB-UHFFFAOYSA-N |
PubChem CID | 81770 |
IUPAC Name | 2,2-dimethylpropane-1,3-diamine |
SMILES | CC(C)(CN)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11935191
|
Thermo Scientific Acros
210510500 |
50 g | Glass bottle |
£87.23
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11945191
|
Thermo Scientific Acros
210512500 |
250 g | Glass bottle |
£281.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7328-91-8 | |
102.18 | |
81770 | |
CC(C)(CN)CN |
C5H14N2 | |
DDHUNHGZUHZNKB-UHFFFAOYSA-N | |
2,2-dimethylpropane-1,3-diamine |
Specifications
7328-91-8 | |
100.0 | |
Colorless | |
152.0°C to 154.0°C | |
Authentic | |
C5H14N2 | |
1.4566 | |
0.85 | |
Solubility in water: miscible | |
CC(C)(CN)CN | |
102.18 | |
102.18 | |
Low Melting Solid |
98.5 | |
29.0°C to 31.0°C | |
0.8500g/mL | |
47°C | |
98.5% min. (GC) | |
H2NCH2C(CH3)2CH2NH2 | |
04,III,595 | |
2,2-dimethyl-1,3-propanediamine, neopentyldiamine, 1,3-propanediamine, 2,2-dimethyl, neopentanediamine, 1,3-diamino-2,2-dimethylpropane, 2,2-dimethyltrimethylenediamine, 1,3-propanediamine,2,2-dimethyl, neopentylenediamine, 2,3-propanediamine, acmc-209oqg | |
DDHUNHGZUHZNKB-UHFFFAOYSA-N | |
2,2-dimethylpropane-1,3-diamine | |
81770 | |
99% | |
2, 2-Dimethyl-1, 3-propanediamine |
Safety and Handling
GHS H Statement
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 230-819-
- 2,2-Dimethyl-1,3-propanediamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only