missing translation for 'onlineSavingsMsg'
Learn More
2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 10783043

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10783043
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10783043

Brand: Thermo Scientific Maybridge CC27201DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 42327-95-7
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD03426826
InChI Key AVVACPBRCFXZMR-UHFFFAOYSA-N
Synonym 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carboxylic acid, 2, 3-dihydro-2, 2-dimethylbenzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carboxylic acid
PubChem CID 2795469
IUPAC Name 2,2-dimethyl-3H-1-benzofuran-7-carboxylic acid
SMILES CC1(CC2=CC=CC(=C2O1)C(=O)O)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.