missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylcyclohexanone, Thermo Scientific Chemicals

Product Code. 10051074
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-47-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.2
InChI Key KNSPBSQWRKKAPI-UHFFFAOYSA-N
Synonym 2,2-dimethylcyclohexanone, cyclohexanone, 2,2-dimethyl, dimethylcyclohexanone, cyclohexanone, dimethyl, 6,6-dimethylcyclohexanone, cyclohexanone,2-dimethyl, acmc-20a0u6, 2,2-dimethyl-cyclohexanone, cyclohexanone,2,2-dimethyl, 2,2-dimethyl-1-cyclohexanone
PubChem CID 136929
IUPAC Name 2,2-dimethylcyclohexan-1-one
SMILES CC1(CCCCC1=O)C

Specifications

Melting Point -20.0°C
Color Colorless to Yellow
Density 0.9120g/mL
Boiling Point 169.0°C to 170.0°C (0.8 mmHg)
Flash Point 50°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2C6H8(=O)
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.912
Formula Weight 126.2
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 2, 2-Dimethylcyclohexanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2-Dimethylcyclohexanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'