Learn More
2,2-Dimethylcyclopentanone, 97%, Thermo Scientific Chemicals
£44.12 - £134.67
Chemical Identifiers
CAS | 4541-32-6 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.17 |
MDL Number | MFCD00049164 |
InChI Key | FTGZMZBYOHMEPS-UHFFFAOYSA-N |
Synonym | 2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one |
PubChem CID | 138286 |
IUPAC Name | 2,2-dimethylcyclopentan-1-one |
SMILES | CC1(CCCC1=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10423274
|
Thermo Scientific Acros
399400010 |
1 g | Glass bottle |
£44.12
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10433274
|
Thermo Scientific Acros
399400050 |
5 g | Glass bottle |
£134.67
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
4541-32-6 | |
112.17 | |
FTGZMZBYOHMEPS-UHFFFAOYSA-N | |
138286 | |
CC1(CCCC1=O)C |
C7H12O | |
MFCD00049164 | |
2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one | |
2,2-dimethylcyclopentan-1-one |
Especificaciones
4541-32-6 | |
100.0 | |
0.8900g/mL | |
32°C | |
96% min. (GC) | |
(CH3)2C5H6(=O) | |
MFCD00049164 | |
2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one | |
CC1(CCCC1=O)C | |
112.17 | |
112.17 | |
Solid |
96.0 | |
White to Yellow | |
143°C to 145°C | |
Authentic | |
C7H12O | |
1.4320 to 1.4340 (20°C, 589nm) | |
0.89 | |
FTGZMZBYOHMEPS-UHFFFAOYSA-N | |
2,2-dimethylcyclopentan-1-one | |
138286 | |
97% | |
2, 2-Dimethylcyclopentanone |
Seguridad y manipulación
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 2,2-Dimethylcyclopentanone
Palabra de advertencia
- Warning
Clasificación
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
Indicaciones de peligro
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H226-Flammable liquid and vapour.
Consejos de prudencia
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only