missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylpropanimidamide hydrochloride, 97%, Thermo Scientific™

Product Code. 10215464
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 18202-73-8
Molecular Formula C5H13ClN2
Molecular Weight (g/mol) 136.623
MDL Number MFCD00051988
InChI Key ARDGQYVTLGUJII-UHFFFAOYSA-N
Synonym 2,2-dimethylpropanimidamide hydrochloride, pivalamidine hydrochloride, pivalimidamide hydrochloride, tert-butylcarbamidine hydrochloride, 2,2-dimethylpropionamidine hydrochloride, 2,2-dimethyl-propionamidine hydrochloride, 2,2-dimethylpropionamidine hcl, t-butylcarbamidine hydrochloride, 2,2-dimethylpropanamidine hydrochloride, 2,2,2-trimethylacetamidine hydrochloride
PubChem CID 2781880
IUPAC Name 2,2-dimethylpropanimidamide;hydrochloride
SMILES CC(C)(C)C(=N)N.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CC(=NH)NH2·HCl
Quantity 10 g
Formula Weight 136.62
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethylpropanimidamide hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.