missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethylpropanimidamide hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)3CC(=NH)NH2·HCl |
Quantity | 25 g |
Formula Weight | 136.62 |
Percent Purity | 97% |
Chemical Name or Material | 2, 2-Dimethylpropanimidamide hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'