Learn More
2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals
£39.30 - £450.00
Chemical Identifiers
CAS | 2127-03-9 |
---|---|
Molecular Formula | C10H8N2S2 |
Molecular Weight (g/mol) | 220.32 |
MDL Number | MFCD00006287 |
InChI Key | HAXFWIACAGNFHA-UHFFFAOYSA-N |
Synonym | 2, 2';-Dipyridyl disulfide |
PubChem CID | 129317592 |
IUPAC Name | 9-[(3aS,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one |
SMILES | S(SC1=CC=CC=N1)C1=CC=CC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10092010
|
Thermo Scientific Acros
162230050 |
5 g | Glass bottle |
£39.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10524101
|
Thermo Scientific Acros
162230250 |
25 g | Glass bottle |
£133.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10781994
|
Thermo Scientific Acros
162231000 |
100 g | Glass bottle |
£450.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2127-03-9 | |
220.32 | |
HAXFWIACAGNFHA-UHFFFAOYSA-N | |
129317592 | |
S(SC1=CC=CC=N1)C1=CC=CC=N1 |
C10H8N2S2 | |
MFCD00006287 | |
2, 2';-Dipyridyl disulfide | |
9-[(3aS,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one |
Specifications
56°C to 60°C | |
Colorless to Tan | |
100.0 | |
97.5% min. (GC) | |
(C5H4N)S-S(C5H4N) | |
21, 45 | |
210°C | |
Solubility in water: 5g/L (20°C). Other solubilities: slightly soluble in methanol,acetone,toluene and,isopropanol,practically insoluble in heptane and cyclohexane | |
S(SC1=CC=CC=N1)C1=CC=CC=N1 | |
220.32 | |
220.32 | |
Authentic | |
2,2′-Dithiodipyridine |
2127-03-9 | |
97.5 | |
Liquid | |
C10H8N2S2 | |
MFCD00006287 | |
06,246; 13,156 | |
2, 2';-Dipyridyl disulfide | |
HAXFWIACAGNFHA-UHFFFAOYSA-N | |
9-[(3aS,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one | |
129317592 | |
98% | |
(5% soln. in methanol) clear |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 218-343-1
- 2,2'-Dithiodipyridine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only