missing translation for 'onlineSavingsMsg'
Learn More

(+)-2,2'-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline], Thermo Scientific Chemicals

Product Code. 11976831
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 150529-93-4
Packaging Glass bottle
Quantity 1 g
Molecular Formula C21H22N2O2
MDL Number MFCD00192244
Merck Index 15, 1297
InChI Key JTNVCJCSECAMLD-UHFFFAOYNA-N
SMILES CC(C)(C1=NC(CO1)C1=CC=CC=C1)C1=NC(CO1)C1=CC=CC=C1
IUPAC Name (4R)-4-phenyl-2-[2-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]propan-2-yl]-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol) 334.42
PubChem CID 676390
Formula Weight 334.42
Melting Point 56.0°C to 58.0°C
Chemical Name or Material (+)-2,2′-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]
Show More Show Less
compliance-icons
Product Identifier
  • (+)-2,2'-Isopropylidenebis¢(4R)-4-phenyl-2-oxazoline!
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.