missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Methoxyethoxy)ethanol, 99%, Thermo Scientific Chemicals
£28.60 - £172.00
Chemical Identifiers
CAS | 111-77-3 |
---|---|
Molecular Formula | C5H12O3 |
Molecular Weight (g/mol) | 120.15 |
MDL Number | MFCD00002871,MFCD00084416 |
InChI Key | SBASXUCJHJRPEV-UHFFFAOYSA-N |
Synonym | 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm |
PubChem CID | 8134 |
ChEBI | CHEBI:44836 |
IUPAC Name | 2-(2-methoxyethoxy)ethanol |
SMILES | COCCOCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10041590
|
Thermo Scientific Acros
149370010 |
1 L | Glass Bottle |
£28.60
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10051590
|
Thermo Scientific Acros
149370025 |
2.5 L | Glass Bottle |
£52.50
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984731
|
Thermo Scientific Acros
149370100 |
10 L | Plastic drum |
£172.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-77-3 | |
120.15 | |
SBASXUCJHJRPEV-UHFFFAOYSA-N | |
8134 | |
2-(2-methoxyethoxy)ethanol |
C5H12O3 | |
MFCD00002871,MFCD00084416 | |
2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm | |
CHEBI:44836 | |
COCCOCCO |
Specifications
111-77-3 | |
1.0100g/mL | |
83°C | |
98.5% min. (GC) | |
CH3OCH2CH2OCH2CH2OH | |
MFCD00002871,MFCD00084416 | |
15,3136 | |
2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm | |
SBASXUCJHJRPEV-UHFFFAOYSA-N | |
2-(2-methoxyethoxy)ethanol | |
8134 | |
3.9 mPa.s (20°C) | |
99% | |
2-(2-Methoxyethoxy)ethanol |
-70.0°C | |
194.0°C | |
Authentic | |
C5H12O3 | |
1.4250 to 1.4270 | |
01,IV,2392 | |
1.01 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol,glycerol,ether,acetone,and dmf | |
COCCOCCO | |
120.15 | |
CHEBI:44836 | |
120.15 | |
Liquid |
Safety and Handling
GHS H Statement
Suspected of damaging the unborn child.
GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-906-6
RUO – Research Use Only