missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Methyl-1,3-thiazol-4-yl)ethanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 143.21 |
Chemical Name or Material | 2-(2-Methyl-1, 3-thiazol-4-yl)ethanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'