missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Naphthyl)acetyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 204.66 |
Percent Purity | 97% |
Chemical Name or Material | 2-(2-Naphthyl)acetyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'