missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
10 g
25 g
Description
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.
Solubility
Soluble in acetone, methanol, water.
Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.
Solubility
Soluble in acetone, methanol, water.
Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 125°C to 126°C (decomposition) |
Color | White |
Quantity | 5 g |
Beilstein | 84382 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in acetone,methanol,water. |
Formula Weight | 260.28 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'