missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Tribromothiophene, 97%, Thermo Scientific™

Product Code. 11306726
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3141-25-1
Molecular Formula C4HBr3S
Molecular Weight (g/mol) 320.82
MDL Number MFCD00041210
InChI Key ZDXQFDMBZUQHOM-UHFFFAOYSA-N
PubChem CID 76593
IUPAC Name 2,3,4-tribromothiophene
SMILES BrC1=C(Br)C(Br)=CS1

Specifications

Melting Point 43°C to 45°C
Boiling Point 150°C to 152°C (20 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 111770
Synonym tribromothiophene, thiophene, 2,3,4-tribromo, pubchem7738, acmc-1bnbq, 3,4,5-tribromothiophene, thiophene,2,3,4-tribromo, buttpark 29\09-41
Formula Weight 320.84
Percent Purity 97%
Chemical Name or Material 2,3,4-Tribromothiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'