missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluoroanisole, 97+%, Thermo Scientific™

Product Code. 11456926
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203245-16-3
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00042396
InChI Key LIVNSQZJDRPZKP-UHFFFAOYSA-N
Synonym 2,3,4-trifluoroanisole, benzene, 1,2,3-trifluoro-4-methoxy-9ci, benzene, 1,2,3-trifluoro-4-methoxy, acmc-20alqi, methyl 2,3,4-trifluorophenyl ether, 1,2,3-trifluoro-4-methoxy-benzene, 2,3,4-trifluorophenol, methyl ether, 1-methoxy-2,3,4-trifluorobenzene
PubChem CID 2776942
IUPAC Name 1,2,3-trifluoro-4-methoxybenzene
SMILES COC1=C(C(=C(C=C1)F)F)F

Specifications

Flash Point 61°C (142°F)
Quantity 5 g
Formula Weight 162.11
Percent Purity ≥97%
Chemical Name or Material 2,3,4-Trifluoroanisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'