missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 95%, Thermo Scientific Chemicals

Product Code. 10460151
Click to view available options
Quantity:
5 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29261-33-4
Melting Point 291.0°C
Assay Percent Range 94% min
Packaging Glass bottle
Molecular Formula C12F4N4
Quantity 5 mg
Synonym 2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 2,2'-perfluorocyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tcnqf4, 2,3,5,6-tetrafluorotetracyanoquinodimethane, f4-tcnq, tetrafluorotetracyanoquinodimethane, 2-4-dicyanomethylidene-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene propanedinitrile, f4tcnq, tetrafluorotetracyanoquinodimethane anion radical, tetrafluorotetracyanoquinodimethane purifiedbysubl
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane,, acetic acid
InChI Key IXHWGNYCZPISET-UHFFFAOYSA-N
SMILES C(#N)C(=C1C(=C(C(=C(C#N)C#N)C(=C1F)F)F)F)C#N
IUPAC Name 2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Molecular Weight (g/mol) 276.16
PubChem CID 2733307
Formula Weight 276.16
Percent Purity 95%
Chemical Name or Material 2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'