Learn More
2,3,5-Collidine, 99%, Thermo Scientific Chemicals
£23.90 - £133.00
Chemical Identifiers
CAS | 695-98-7 |
---|---|
Molecular Formula | C8H11N |
Molecular Weight (g/mol) | 121.183 |
MDL Number | MFCD00126840 |
InChI Key | GFYHSKONPJXCDE-UHFFFAOYSA-N |
Synonym | 2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f |
PubChem CID | 12759 |
IUPAC Name | 2,3,5-trimethylpyridine |
SMILES | CC1=CC(=C(N=C1)C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11455547
|
Thermo Scientific Alfa Aesar
B23537.06 |
5 g |
£23.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465547
|
Thermo Scientific Alfa Aesar
B23537.14 |
25 g |
£48.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475547
|
Thermo Scientific Alfa Aesar
B23537.22 |
100 g |
£133.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
Chemical Identifiers
695-98-7 | |
121.183 | |
GFYHSKONPJXCDE-UHFFFAOYSA-N | |
12759 | |
CC1=CC(=C(N=C1)C)C |
C8H11N | |
MFCD00126840 | |
2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f | |
2,3,5-trimethylpyridine |
Specifications
695-98-7 | |
182°C to 184°C | |
Unpleasant | |
1.5075 | |
UN2810 | |
2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f | |
GFYHSKONPJXCDE-UHFFFAOYSA-N | |
2,3,5-trimethylpyridine | |
12759 | |
99% |
0.935 | |
74°C (165°F) | |
C8H11N | |
MFCD00126840 | |
108832 | |
Slightly soluble in water. | |
CC1=CC(=C(N=C1)C)C | |
121.183 | |
121.18 | |
2,3,5-Collidine |
Safety and Handling
GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'einecsNumber' : 211-786-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only