missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[3,5-di(tert-butyl)-4-hydroxyphenyl]acetic acid, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Specifications
Quantity | 10 g |
Formula Weight | 264.36 |
Percent Purity | 97% |
Chemical Name or Material | 2-[3, 5-di(tert-butyl)-4-hydroxyphenyl]acetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'