missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-difluorophenyl)acetic acid, Thermo Scientific™

£20.70 - £137.00

Chemical Identifiers

CAS 105184-38-1
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
InChI Key IGGNSAVLXJKCNH-UHFFFAOYSA-N
Synonym 3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
PubChem CID 145424
IUPAC Name 2-(3,5-difluorophenyl)acetic acid
SMILES C1=C(C=C(C=C1F)F)CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10066992
View Documents
Thermo Scientific Maybridge
SB01687EA
10 g
£20.70
10g
Please to purchase this item. Need a web account? Register with us today!
10082364
View Documents
Thermo Scientific Maybridge
SB01687FA
100 g
£137.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

105184-38-1
172.131
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
2-(3,5-difluorophenyl)acetic acid
C8H6F2O2
IGGNSAVLXJKCNH-UHFFFAOYSA-N
145424
C1=C(C=C(C=C1F)F)CC(=O)O

Specifications

105184-38-1
C8H6F2O2
IGGNSAVLXJKCNH-UHFFFAOYSA-N
2-(3,5-difluorophenyl)acetic acid
145424
97%
97%
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
C1=C(C=C(C=C1F)F)CC(=O)O
172.131
172.13
2-(3, 5-Difluorophenyl)acetic acid
Videos
SDS
Documents

Documents

Product Certifications