missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Difluorophenyl)acetonitrile, 97%, Thermo Scientific™

Product Code. 10101232
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 122376-76-5
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD00061278
InChI Key OBMYMTSBABEPIB-UHFFFAOYSA-N
Synonym 3,5-difluorophenylacetonitrile, 2-3,5-difluorophenyl acetonitrile, 3,5-difluorobenzyl cyanide, 3,5-difluorobenzylcyanide, benzeneacetonitrile, 3,5-difluoro, 3,5-difluorophenyl acetonitrile, 2-3,5-difluorophenyl ethanenitrile, pubchem1588, acmc-1bxc0
PubChem CID 518565
IUPAC Name 2-(3,5-difluorophenyl)acetonitrile
SMILES C1=C(C=C(C=C1F)F)CC#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula F2C6H3CH2CN
Quantity 10 g
Formula Weight 153.13
Percent Purity 97%
Chemical Name or Material 2-(3, 5-Difluorophenyl)acetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'