missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Difluorophenyl)ethanoyl chloride, 97%, Thermo Scientific™

Product Code. 10170562
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 157033-24-4
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD00275597
InChI Key WMVPARKLYWQLNN-UHFFFAOYSA-N
PubChem CID 2782723
IUPAC Name 2-(3,5-difluorophenyl)acetyl chloride
SMILES C1=C(C=C(C=C1F)F)CC(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula F2C6H3CH2C(O)Cl
Quantity 250 mg
Synonym 2-3,5-difluorophenyl ethanoyl chloride, 2-3,5-difluorophenyl acetyl chloride, 3,5-difluorophenylacetyl chloride, benzeneacetyl chloride, 3,5-difluoro, benzeneacetyl chloride,3,5-difluoro, acmc-20dux9, 3.5-difluorophenylacetyl chloride, 3,5-difluorophenyl acetyl chloride, 3,5-difluorophenyl-acetyl chloride, 3,5-difluoro-phenyl-acetyl chloride
Formula Weight 190.58
Percent Purity 97%
Chemical Name or Material 2-(3, 5-Difluorophenyl)ethanoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'