missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3,5-Difluorophenyl)ethanoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | F2C6H3CH2C(O)Cl |
Quantity | 1 g |
Synonym | 2-3,5-difluorophenyl ethanoyl chloride, 2-3,5-difluorophenyl acetyl chloride, 3,5-difluorophenylacetyl chloride, benzeneacetyl chloride, 3,5-difluoro, benzeneacetyl chloride,3,5-difluoro, acmc-20dux9, 3.5-difluorophenylacetyl chloride, 3,5-difluorophenyl acetyl chloride, 3,5-difluorophenyl-acetyl chloride, 3,5-difluoro-phenyl-acetyl chloride |
Formula Weight | 190.58 |
Percent Purity | 97% |
Chemical Name or Material | 2-(3, 5-Difluorophenyl)ethanoyl chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction