missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid, 97%, Thermo Scientific™

£29.90 - £41.80

Chemical Identifiers

CAS 321309-43-7
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD02682016
InChI Key JNHKHFADRGJMJS-UHFFFAOYSA-N
Synonym 2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
PubChem CID 2776446
IUPAC Name 2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
SMILES CC1=C(C(C)=NN1)C1=CC=CC=C1C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10602633
SDS View Documents
Thermo Scientific Maybridge
CC15701CB
250 mg Amber Glass Bottle
£29.90
250mg
Please to purchase this item. Need a web account? Register with us today!
10772273
SDS View Documents
Thermo Scientific Maybridge
CC15701DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.