missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-beta-D-arabinofuranose, Thermo Scientific Chemicals

Product Code. 15468379
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

An arabinofuranose derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An arabinofuranose derivative.

Solubility
Soluble in methanol

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60933-68-8
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD00047076
InChI Key NAQUAXSCBJPECG-FXSWLTOZSA-N
Synonym 2,3,5-tri-o-benzyl-beta-d-arabinofuranose, 2,3,5-tri-o-benzyl-b-d-arabinofuranose, 2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2-o,3-o,5-o-tribenzyl-beta-d-arabinofuranose, 2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
PubChem CID 11729473
IUPAC Name (2R,3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in methanol
Formula Weight 420.5
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'