missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trifluorophenylacetic acid, 97%, Thermo Scientific™

Product Code. 11316969
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132992-28-0
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00083547
InChI Key HHVKNKDKNBABCU-UHFFFAOYSA-N
Synonym 2,3,5-trifluorophenylacetic acid, 2-2,3,5-trifluorophenyl acetic acid, 2,3,5-trifluorophenyl acetic acid, benzeneacetic acid, 2,3,5-trifluoro, pubchem4205, intermediates-zcf02025, acmc-1c03i, 2,3,5-trifluorobenzeneacetic acid, 2-2,3,5-trifluorophenyl aceticacid
PubChem CID 2777946
IUPAC Name 2-(2,3,5-trifluorophenyl)acetic acid
SMILES C1=C(C=C(C(=C1F)F)CC(=O)O)F

Specifications

Quantity 1 g
Formula Weight 190.12
Percent Purity 97%
Chemical Name or Material 2,3,5-Trifluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'