missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethoxybenzeneboronic acid, 95%, Thermo Scientific™

Product Code. 15419957
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-67-1
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD03411944
InChI Key PBJYOPCURLYSAG-UHFFFAOYSA-N
Synonym 2,3,6-trimethoxybenzeneboronic acid, 2,3,6-trimethoxyphenyl boronic acid, 2,3,6-trimethoxy benzeneboronic acid, acmc-1ctjt, boronic acid, 2,3,6-trimethoxyphenyl-9ci, boronic acid, 2,3,6-trimethoxyphenyl
PubChem CID 2773583
IUPAC Name (2,3,6-trimethoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1OC)OC)OC)(O)O

Specifications

Melting Point 34°C to 40°C
Quantity 1 g
Formula Weight 212.01
Percent Purity 95%
Chemical Name or Material 2,3,6-Trimethoxybenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'