missing translation for 'onlineSavingsMsg'
Learn More
2,3,6-Trimethoxybenzeneboronic acid, 95%, Thermo Scientific™ Product Code.: 15429957

2,3,6-Trimethoxybenzeneboronic acid, 95%, Thermo Scientific™

Product Code. 15429957
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15429957

Brand: Thermo Scientific Alfa Aesar H53197.MD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-67-1
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD03411944
InChI Key PBJYOPCURLYSAG-UHFFFAOYSA-N
Synonym 2,3,6-trimethoxybenzeneboronic acid, 2,3,6-trimethoxyphenyl boronic acid, 2,3,6-trimethoxy benzeneboronic acid, acmc-1ctjt, boronic acid, 2,3,6-trimethoxyphenyl-9ci, boronic acid, 2,3,6-trimethoxyphenyl
PubChem CID 2773583
IUPAC Name (2,3,6-trimethoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1OC)OC)OC)(O)O

Specifications

Melting Point 34°C to 40°C
Quantity 250 mg
Formula Weight 212.01
Percent Purity 95%
Chemical Name or Material 2,3,6-Trimethoxybenzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE02Y052 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3,6-Trimethoxybenzeneboronic acid, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.