missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,6-Trimethylphenol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.
Solubility
Soluble in methanol.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.
Solubility
Soluble in methanol.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 59°C to 62°C |
Density | 0.94 |
Boiling Point | 215°C |
Flash Point | 100°C (212°F) |
Quantity | 25 g |
UN Number | UN2430 |
Beilstein | 2042207 |
Solubility Information | Soluble in methanol. |
Formula Weight | 136.19 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'