missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%, Thermo Scientific Chemicals

£48.24 - £166.83

Chemical Identifiers

CAS 124215-44-7
Molecular Formula C9H18BBrO2
Molecular Weight (g/mol) 248.96
MDL Number MFCD10567053
InChI Key CHQXFJUKMDJWHO-UHFFFAOYSA-N
Synonym 2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane
PubChem CID 14836669
IUPAC Name 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(CCCBr)OC1(C)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10676840
View Documents
Thermo Scientific Acros
433930010
1 g Glass bottle
£48.24
1g
Please to purchase this item. Need a web account? Register with us today!
10525490
View Documents
Thermo Scientific Acros
433930050
5 g Glass bottle
£166.83
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124215-44-7
248.96
CHQXFJUKMDJWHO-UHFFFAOYSA-N
14836669
CC1(C)OB(CCCBr)OC1(C)C
C9H18BBrO2
MFCD10567053
2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane
2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

124215-44-7
100.0
1.1770g/mL
Authentic
C9H18BBrO2
1.177
CHQXFJUKMDJWHO-UHFFFAOYSA-N
2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
14836669
97%
2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
96.0
Colorless to Yellow
61°C to 64°C (0.4 mmHg)
96% min. (GC)
MFCD10567053
2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane
CC1(C)OB(CCCBr)OC1(C)C
248.96
248.95
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only