missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals

Product Code. 11975651
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-71-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00002116
InChI Key FSEUPUDHEBLWJY-HWKANZROSA-N
Synonym 2,3-butanedione monoxime, diacetyl monoxime, diacetylmonoxime, biacetyl monoxime, biacetyl monooxime, diacetyl monooxime, diacetylmonooxime, 2-oximino-3-butanone, 3-oximino-2-butanone, 2,3-butanedione-2-monoxime
PubChem CID 6409633
ChEBI CHEBI:4480
IUPAC Name (3E)-3-hydroxyiminobutan-2-one
SMILES CC(=O)C(\C)=N\O

Specifications

Melting Point 74°C to 76°C
Color White
Boiling Point 185°C to 186°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(=NOH)COCH3
Quantity 2.5 kg
Beilstein 01,772
Solubility Information (5% in water) Clear to slightly hazy
Formula Weight 101.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Butanedione monoxime
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'