missing translation for 'onlineSavingsMsg'
Learn More
2-[3-(Chloromethyl)piperidino]-6-methylpyrazine, 97%, Thermo Scientific™ Product Code.: 10738265

2-[3-(Chloromethyl)piperidino]-6-methylpyrazine, 97%, Thermo Scientific™

Product Code. 10738265
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10738265

Brand: Thermo Scientific Maybridge CC59024CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 937795-92-1
Molecular Formula C11H16ClN3
Molecular Weight (g/mol) 225.72
MDL Number MFCD09879918
InChI Key DTVYKEHYUMEUFH-UHFFFAOYNA-N
Synonym 2-3-chloromethyl piperidin-1-yl-6-methylpyrazine, 2-3-chloromethyl piperidino-6-methylpyrazine, 2-3-chloromethyl piperidyl-6-methylpyrazine
PubChem CID 24229601
IUPAC Name 2-[3-(chloromethyl)piperidin-1-yl]-6-methylpyrazine
SMILES CC1=CN=CC(=N1)N1CCCC(CCl)C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 225.72
Percent Purity 97%
Chemical Name or Material 2-[3-(Chloromethyl)piperidino]-6-methylpyrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-[3-(Chloromethyl)piperidino]-6-methylpyrazine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.