missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11414637
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2973-75-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00016978
InChI Key WKLKGSHBXNPUDU-UHFFFAOYSA-N
Synonym 5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
PubChem CID 520452
IUPAC Name 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=C(Br)C(Br)=C1O

Specifications

Melting Point 213°C to 217°C
Quantity 5 g
Beilstein 1968576
Sensitivity Air Sensitive
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material 2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'