Learn More
2,3-Dibromosuccinic acid, 98%, Thermo Scientific™
£76.45
Chemical Identifiers
CAS | 526-78-3 |
---|---|
Molecular Formula | C4H4Br2O4 |
Molecular Weight (g/mol) | 275.88 |
MDL Number | MFCD00004209 |
InChI Key | FJWGRXKOBIVTFA-UHFFFAOYSA-N |
Synonym | 2,3-dibromosuccinic acid, butanedioic acid, 2,3-dibromo, 2,3-dibromosuccinicacid, sym.-dibromosuccinic acid, acmc-20a6bk, succinic acid,3-dibromo, 2,3-dibromo succinic acid, butanedioic acid,3-dibromo, 2,3-dibromosuccinic acid #, 2,3-dibromo-butanedioic acid |
PubChem CID | 95324 |
IUPAC Name | 2,3-dibromobutanedioic acid |
SMILES | C(C(C(=O)O)Br)(C(=O)O)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436778
|
Thermo Scientific Alfa Aesar
H60306.22 |
100 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
526-78-3 | |
275.88 | |
FJWGRXKOBIVTFA-UHFFFAOYSA-N | |
95324 | |
C(C(C(=O)O)Br)(C(=O)O)Br |
C4H4Br2O4 | |
MFCD00004209 | |
2,3-dibromosuccinic acid, butanedioic acid, 2,3-dibromo, 2,3-dibromosuccinicacid, sym.-dibromosuccinic acid, acmc-20a6bk, succinic acid,3-dibromo, 2,3-dibromo succinic acid, butanedioic acid,3-dibromo, 2,3-dibromosuccinic acid #, 2,3-dibromo-butanedioic acid | |
2,3-dibromobutanedioic acid |
Specifications
526-78-3 | |
C4H4Br2O4 | |
MFCD00004209 | |
2,3-dibromosuccinic acid, butanedioic acid, 2,3-dibromo, 2,3-dibromosuccinicacid, sym.-dibromosuccinic acid, acmc-20a6bk, succinic acid,3-dibromo, 2,3-dibromo succinic acid, butanedioic acid,3-dibromo, 2,3-dibromosuccinic acid #, 2,3-dibromo-butanedioic acid | |
C(C(C(=O)O)Br)(C(=O)O)Br | |
275.88 | |
275.88 | |
Solid |
274°C | |
HOOCCHBrCHBrCOOH | |
UN3261 | |
FJWGRXKOBIVTFA-UHFFFAOYSA-N | |
2,3-dibromobutanedioic acid | |
95324 | |
98% | |
2,3-Dibromosuccinic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 208-396-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures