Learn More
2,3-Dichloro-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals
£19.40 - £104.00
Chemical Identifiers
CAS | 117-80-6 |
---|---|
Molecular Formula | C10H4Cl2O2 |
Molecular Weight (g/mol) | 227.04 |
MDL Number | MFCD00001677 |
InChI Key | SVPKNMBRVBMTLB-UHFFFAOYSA-N |
Synonym | 2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone |
PubChem CID | 8342 |
SMILES | ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474110
|
Thermo Scientific Alfa Aesar
A13378.14 |
25 g |
£19.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465363
|
Thermo Scientific Alfa Aesar
A13378.22 |
100 g |
£28.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475363
|
Thermo Scientific Alfa Aesar
A13378.36 |
500 g |
£104.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Use in textiles, as organic catalyst, as chemical intermediate, as additive in dye binders. Also used as an important catalyst to produce 3,3-dichlorobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUse as seed disinfectant, textiles, as organic catalyst, as chemical intermediate, as additive in dye binders, as antifouling agent. Also used to control blue-green algae in ponds, lakes, and swimming pools. Also used as an important catalyst to produce 3,3-Dichloro benzine.
Solubility
Soluble in water (0.008g/L). Soluble in hot toluene almost transparency.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
117-80-6 | |
227.04 | |
SVPKNMBRVBMTLB-UHFFFAOYSA-N | |
8342 |
C10H4Cl2O2 | |
MFCD00001677 | |
2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone | |
ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O |
Specifications
117-80-6 | |
275°C (2 mmHg) | |
MFCD00001677 | |
1073511 | |
2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone | |
SVPKNMBRVBMTLB-UHFFFAOYSA-N | |
2,3-dichloronaphthalene-1,4-dione | |
8342 | |
98% |
193°C to 196°C (sublimation) | |
C10H4Cl2O2 | |
UN2811 | |
14,3045 | |
Soluble in water (0.008g/L). Soluble in hot toluene almost transparency. | |
ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O | |
227.04 | |
227.05 | |
2,3-Dichloro-1,4-naphthoquinone |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319-H500
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 204-210-5
missing translation for 'rtecsNumber' : QL7525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2,3-Dichloro-1,4-naphthoquinone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 2,3-Dichloro-1,4-naphthoquinone
RUO – Research Use Only