Learn More
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals
£29.50 - £341.00
Chemical Identifiers
CAS | 84-58-2 |
---|---|
Molecular Formula | C8Cl2N2O2 |
Molecular Weight (g/mol) | 227 |
MDL Number | MFCD00001593 |
InChI Key | HZNVUJQVZSTENZ-UHFFFAOYSA-N |
Synonym | 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile |
PubChem CID | 6775 |
IUPAC Name | 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile |
SMILES | C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453844
|
Thermo Scientific Alfa Aesar
A11879.09 |
10 g |
£29.50
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463844
|
Thermo Scientific Alfa Aesar
A11879.18 |
50 g |
£93.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473844
|
Thermo Scientific Alfa Aesar
A11879.30 |
250 g |
£341.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone is used as a reagent for oxidative couplings and cyclization reactions and dehydrogenation of alcohols, phenols and steroid ketones. It is also used in the synthesis of 1,2-benzisoxazoles. It is a useful electron-transfer reagent for synthesis of quinolines from imines and alkynes or alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dichloro-5,6-dicyano-1,4-benzoquinone is used as a reagent for oxidative couplings and cyclization reactions and dehydrogenation of alcohols, phenols and steroid ketones. It is also used in the synthesis of 1,2-benzisoxazoles. It is a useful electron-transfer reagent for synthesis of quinolines from imines and alkynes or alkenes.
Solubility
Soluble in benzene, methanol, acetic acid and dioxane.Slightly soluble in chloroform,, dichloromethane.
Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Chemical Identifiers
84-58-2 | |
227 | |
HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
6775 | |
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N |
C8Cl2N2O2 | |
MFCD00001593 | |
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile | |
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile |
Specifications
84-58-2 | |
C8Cl2N2O2 | |
UN3439 | |
Moisture sensitive | |
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile | |
HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile | |
6775 | |
≥98% |
214°C to 218°C | |
MFCD00001593 | |
747939 | |
14,3063 | |
Soluble in benzene,methanol,acetic acid and dioxane.Slightly soluble in chloroform,,dichloromethane. | |
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N | |
227 | |
227.01 | |
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P280-P301+P310-P305+P351+P338-P310-P330-P501c
H301-H318
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 201-542-2
missing translation for 'rtecsNumber' : GU4825000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
RUO – Research Use Only