Learn More
2,3-Dichloroaniline, 99%, Thermo Scientific Chemicals
£64.30 - £192.00
Chemical Identifiers
CAS | 608-27-5 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.013 |
MDL Number | MFCD00007657 |
InChI Key | BRPSAOUFIJSKOT-UHFFFAOYSA-N |
Synonym | benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine |
PubChem CID | 11844 |
ChEBI | CHEBI:46636 |
IUPAC Name | 2,3-dichloroaniline |
SMILES | C1=CC(=C(C(=C1)Cl)Cl)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404330
|
Thermo Scientific Alfa Aesar
A11524.22 |
100 g |
£64.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414330
|
Thermo Scientific Alfa Aesar
A11524.36 |
500 g |
£192.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494320
|
Thermo Scientific Alfa Aesar
A11524.0E |
2500 g | N/A | ||||||
Description
2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.
Solubility
Soluble in ether, alcohol and acetone. Slightly soluble in benzene. Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Chemical Identifiers
608-27-5 | |
162.013 | |
BRPSAOUFIJSKOT-UHFFFAOYSA-N | |
11844 | |
2,3-dichloroaniline |
C6H5Cl2N | |
MFCD00007657 | |
benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine | |
CHEBI:46636 | |
C1=CC(=C(C(=C1)Cl)Cl)N |
Specifications
608-27-5 | |
250°C to 252°C | |
C6H5Cl2N | |
UN3442 | |
benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine | |
C1=CC(=C(C(=C1)Cl)Cl)N | |
162.013 | |
CHEBI:46636 | |
99% |
20°C to 24°C | |
115°C (239°F) | |
MFCD00007657 | |
472027 | |
BRPSAOUFIJSKOT-UHFFFAOYSA-N | |
2,3-dichloroaniline | |
11844 | |
162.02 | |
2,3-Dichloroaniline |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
missing translation for 'rtecsNumber' : CX9862625
missing translation for 'tsca' : TSCA
RUO – Research Use Only