missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

£20.60 - £185.93

Chemical Identifiers

CAS 151169-74-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01075703
InChI Key TYIKXPOMOYDGCS-UHFFFAOYSA-N
Synonym 2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
PubChem CID 2734661
IUPAC Name (2,3-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(Cl)=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479817
SDS View Documents
Thermo Scientific Alfa Aesar
B22781.03
1 g
£20.60
1g
Please to purchase this item. Need a web account? Register with us today!
11489817
SDS View Documents
Thermo Scientific Alfa Aesar
B22781.06
5 g
£61.51
5g
Please to purchase this item. Need a web account? Register with us today!
11499817
SDS View Documents
Thermo Scientific Alfa Aesar
B22781.14
25 g
£185.93
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Dichlorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.