missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11415013
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82417-45-6
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00051844
InChI Key PQODWTNHDKDHIW-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r
PubChem CID 2734222
IUPAC Name 2,3-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 57°C to 62°C
Boiling Point 131°C to 133°C (1 mmHg)
Quantity 100 g
UN Number UN3261
Beilstein 7703966
Sensitivity Moisture sensitive
Formula Weight 245.51
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'