missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzonitrile, 98%

Product Code. 11462557
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11462557

Brand: Thermo Scientific Alfa Aesar A14809.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6574-97-6 | C7H3Cl2N | 172.008 g/mol

2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
It is miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6574-97-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016372
InChI Key OHDYZVVLNPXKDX-UHFFFAOYSA-N
Synonym benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b
PubChem CID 736567
IUPAC Name 2,3-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)Cl)C#N

Specifications

Melting Point 60°C to 65°C
Boiling Point 114°C (6 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 2438753
Solubility Information It is miscible with water.
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.