missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals

£23.30 - £222.00

Chemical Identifiers

CAS 151169-74-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01075703
InChI Key TYIKXPOMOYDGCS-UHFFFAOYSA-N
Synonym 2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
PubChem CID 2734661
IUPAC Name (2,3-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(Cl)=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10591852
missing translation for 'viewDocuments'
Thermo Scientific Acros
367710010
1 g Glass bottle
£23.30
1g
Please to purchase this item. Need a web account? Register with us today!
10792823
missing translation for 'viewDocuments'
Thermo Scientific Acros
367710050
5 g Glass bottle
£62.20
5g
Please to purchase this item. Need a web account? Register with us today!
10667854
missing translation for 'viewDocuments'
Thermo Scientific Acros
367710250
25 g Glass bottle
£222.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

151169-74-3
190.81
TYIKXPOMOYDGCS-UHFFFAOYSA-N
2734661
OB(O)C1=C(Cl)C(Cl)=CC=C1
C6H5BCl2O2
MFCD01075703
2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
(2,3-dichlorophenyl)boronic acid

Specifications

151169-74-3
100.0
White
97%
MFCD01075703
TYIKXPOMOYDGCS-UHFFFAOYSA-N
(2,3-dichlorophenyl)boronic acid
2734661
97%
2, 3-Dichlorophenylboronic acid
96.0
242°C to 244°C
Authentic
C6H5BCl2O2
2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
OB(O)C1=C(Cl)C(Cl)=CC=C1
190.81
190.82
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

Certificates

RUO – Research Use Only