missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methoxy-DL-phenylalanine, 97%, Thermo Scientific™

Product Code. 15465577
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256482-63-9
Molecular Formula C10H11F2NO3
Molecular Weight (g/mol) 231.20
MDL Number MFCD09832370
InChI Key PQCGRBWGXKGWIG-UHFFFAOYNA-N
Synonym 2,3-difluoro-4-methoxy-dl-phenylalanine, 2-amino-3-2,3-difluoro-4-methoxyphenyl propanoic acid, 2,3-difluoro-4-methoxy-dl-phenylalanine, jrd
PubChem CID 46737684
IUPAC Name 2-amino-3-(2,3-difluoro-4-methoxyphenyl)propanoic acid
SMILES COC1=C(F)C(F)=C(CC(N)C(O)=O)C=C1

Specifications

Melting Point ∽260°C (decomposition)
Quantity 1 g
Formula Weight 231.2
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-4-methoxy-DL-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'