missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methoxyaniline, 97%, Thermo Scientific™

Product Code. 11336779
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155020-51-2
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.136
MDL Number MFCD04115986
InChI Key MEETZAMYCFAKKW-UHFFFAOYSA-N
Synonym 2,3-difluoro-p-anisidine, 2, 3-difluoro-4-methoxyaniline, 2,3-difluoro-4-methoxybenzenamine, 2,3-difluoro-4-methoxyphenylamine, benzenamine,2,3-difluoro-4-methoxy
PubChem CID 3798495
IUPAC Name 2,3-difluoro-4-methoxyaniline
SMILES COC1=C(C(=C(C=C1)N)F)F

Specifications

Melting Point 38°C to 40°C
Quantity 5 g
UN Number UN2811
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-4-methoxyaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'