missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methylphenol, 97%, Thermo Scientific™

Product Code. 11396949
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261763-45-5
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD01631651
InChI Key NVYIUITYVMVHJM-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro-4-methyl, acmc-20ajxd, 2,3-difluoro-4-methyl phenol, 2,3-difluoro-4-methyl-phenol, phenol, 2,3-difluoro-4-methyl-9ci
PubChem CID 2774151
IUPAC Name 2,3-difluoro-4-methylphenol
SMILES CC1=C(F)C(F)=C(O)C=C1

Specifications

Melting Point 35°C to 38°C
Quantity 5 g
UN Number UN3261
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-4-methylphenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'