missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxine-2-carboxylic acid, 97%, Thermo Scientific™
£8.15 - £15.00
Chemical Identifiers
CAS | 3663-80-7 |
---|---|
Molecular Formula | C9H7O4 |
Molecular Weight (g/mol) | 179.15 |
MDL Number | MFCD00084888 |
InChI Key | HMBHAQMOBKLWRX-QMMMGPOBSA-M |
Synonym | 1,4-benzodioxan-2-carboxylic acid, 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-2-carboxylic acid, 1,4-benzodioxin-2-carboxylic acid, 2,3-dihydro, 1,4-benzodioxan-2-carboxylicacid, 2rs-2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, rac 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-2-carboxylic acid |
PubChem CID | 2735450 |
IUPAC Name | 2,3-dihydro-1,4-benzodioxine-3-carboxylic acid |
SMILES | [O-]C(=O)[C@@H]1COC2=CC=CC=C2O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10180582
|
Thermo Scientific Maybridge
CC01501CB |
250 mg |
£8.15
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10732283
|
Thermo Scientific Maybridge
CC01501DA |
1 g |
£9.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10016374
|
Thermo Scientific Maybridge
CC01501DE |
5 g |
£15.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
3663-80-7 | |
179.15 | |
HMBHAQMOBKLWRX-QMMMGPOBSA-M | |
2735450 | |
[O-]C(=O)[C@@H]1COC2=CC=CC=C2O1 |
C9H7O4 | |
MFCD00084888 | |
1,4-benzodioxan-2-carboxylic acid, 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-2-carboxylic acid, 1,4-benzodioxin-2-carboxylic acid, 2,3-dihydro, 1,4-benzodioxan-2-carboxylicacid, 2rs-2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, rac 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-2-carboxylic acid | |
2,3-dihydro-1,4-benzodioxine-3-carboxylic acid |
Specifications
3663-80-7 | |
Amber glass bottle | |
MFCD00084888 | |
HMBHAQMOBKLWRX-QMMMGPOBSA-M | |
2,3-dihydro-1,4-benzodioxine-3-carboxylic acid | |
2735450 | |
97% |
97% | |
C9H7O4 | |
1,4-benzodioxan-2-carboxylic acid, 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-2-carboxylic acid, 1,4-benzodioxin-2-carboxylic acid, 2,3-dihydro, 1,4-benzodioxan-2-carboxylicacid, 2rs-2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, rac 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-2-carboxylic acid | |
[O-]C(=O)[C@@H]1COC2=CC=CC=C2O1 | |
179.15 | |
180.16 | |
2, 3-Dihydro-1, 4-benzodioxine-2-carboxylic acid |